Skip to Content
Merck
All Photos(1)

Key Documents

Z509000

Sigma-Aldrich

AldraSORB water trapping packets

volume 25-200 mL, small

Synonym(s):

AldraSORB Packets, Trapping Packets, Water Trapping Packets

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$133.00
25 EA
$493.00

$133.00


Check Cart for Availability


Select a Size

Change View
5 EA
$133.00
25 EA
$493.00

About This Item

UNSPSC Code:
24141516
NACRES:
NB.43

$133.00


Check Cart for Availability

capacity

25-200 mL

volume

25-200 mL

weight

2 g

Looking for similar products? Visit Product Comparison Guide

General description

The AldraSorb packets are made from an ultra-high molecular weight polyethylene, electronic grade, hydrophobic membrane that contains pharmaceutical quality molecular sieves. The full filtration capability of the molecular particulates is maintained and the surrounding solvent is not compromised. Packets are vacuum sealed.

  • Maintains low levels of water impurities for up to 3 months
  • Ideal for solid-phase and solution-phase peptide synthesis
  • Effectively dries solvents to below 50 ppm

Note: These traps are not recommended for solvents having more than 0.05% or 500 ppm of water. They should not be used in acidic solvents.

Legal Information

AldraSORB is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service