Key Documents
Z507865
Round-bottom flask with side port
capacity 10 mL
Select a Size
1 EA
$112.00
$112.00
Check Cart for Availability
Select a Size
Change View
1 EA
$112.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$112.00
Check Cart for Availability
material
borosilicate glass
round bottom flask
single-neck
sterility
non-sterile
feature
closure type Screw cap
packaging
pack of 1
manufacturer/tradename
Ace Glass 9592-37
capacity
10 mL
General description
Single neck flask has threaded <ST/>14/10 joint, and an additional side port with PTFE-faced silicone rubber septum for easy retrieval or injection access. Supplied with holed caps and FETFE® O-rings.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)