Key Documents
Z507237
Supelco
Michel-Miller Swagelok® adapter
NPTF 1/16 in., PTFE, Ace-Thred for #11
Select a Size
1 EA
$130.00
$130.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$130.00
About This Item
UNSPSC Code:
12352200
NACRES:
SB.52
$130.00
Check Cart for Availability
Request a Bulk Order
material
PTFE
manufacturer/tradename
Ace Glass 584444
technique(s)
LPLC: suitable
NPTF
1/16 in.
fitting
Ace-Thred for #11
General description
Use to make Swagelok tubing connections with Ace threaded glassware. One end is Ace-Thred, other is NPTF. Connects tubing to M-M columns. Adapters with #50 Ace-Thred fit Ace large columns.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)