Key Documents
Z506559
Perfektum® one-way stainless steel stopcock
female Luer to male Luer lock
Select a Size
1 EA
$540.00
$540.00
Estimated to ship on14 April 2025
Select a Size
Change View
1 EA
$540.00
About This Item
UNSPSC Code:
41122000
NACRES:
NB.03
$540.00
Estimated to ship on14 April 2025
feature
CE compliant
manufacturer/tradename
Cadence Science 6021SS
parameter
45-60 psi pressure range (this fitting is pressure tested using water)
General description
Made from corrosion-resistant 303 stainless steel.
These adapters have been specifically designed to minimize stripping of plastic Luers when mated with metal Luers.
Manufactured to ISO 594 1/2 standards and meet FDA and CE requirements.
Legal Information
Perfektum is a registered trademark of Cadence Science
Choose from one of the most recent versions:
Certificates of Analysis (COA)