Key Documents
Z408530
Scienceware® Vikem® vinyl-coated lead wire
wire diam. 5/16 in.
Synonym(s):
coil wire, wire spiral, wire vinyl
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
material
lead (coated)
sterility
non-sterile
packaging
pack of 1
manufacturer/tradename
Bel-Art
wire diam.
5/16 in.
weight
176 g
General description
Can be coiled into a helix, spiral, cone, or any irregular shape to fit around flasks and bottle necks to stabilize them in a water bath. 12 in. length. Coated with chemically-resistant red Vikem vinyl.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Vikem is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)