Key Documents
Z408468
Sigma-Aldrich
Scienceware® ROTO-FLO flow indicator
fits tubing, 5/16 - 1/2in., green wheel
Select a Size
1 EA
$53.30
$53.30
Available to ship on23 April 2025Details
Select a Size
Change View
1 EA
$53.30
About This Item
UNSPSC Code:
20121904
NACRES:
NB.51
$53.30
Available to ship on23 April 2025Details
description
barbed ends
feature
fits tubing 5/16 - 1/2in.
manufacturer/tradename
Bel-Art
color
green wheel
General description
Colored paddle wheel spins freely with liquid or gas flow (range: 0.5 to 10 L/min). Made of clear PS. Not for use with corrosive fluids nor at temperatures above 64 °C.
Legal Information
ROTO-FLO is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)