Skip to Content
Merck
All Photos(1)

Key Documents

Z404705

IKA® RW 11 "Lab egg" stirrer

Synonym(s):

lab mixers, laboratory mixer, overhead stirrers

Sign Into View Organizational & Contract Pricing


About This Item

AC/DC input

100 V / 240 V (100-240V 50/60Hz)

parameter

0-2000 rpm speed

H × W × D

89 mm × 86 mm × 175 mm

weight

0.4 kg

Looking for similar products? Visit Product Comparison Guide

General description

IKA® RW 11 Basic "Lab egg" Stirrer

  • Ideal for stirring small quantities (up to 2 L)
  • Borosilicate glass bottom (glass housing resistant to chemicals)
  • Supplied with a vane stirrer (including paddle stirrer R 1001 and extension arm)
  • Viscosity range: 0 to 100 mPas
  • Motor power, consumption: 8 W, output: 1 W

Legal Information

IKA is a registered trademark of IKA-Werke GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service