Key Documents
Z341436
Sigma-Aldrich
Galileo thermometer
Grande, temperature 18-27 °C
About This Item
UNSPSC Code:
41113319
manufacturer/tradename
HB Instruments Grande 250 Celsium
temperature measuring range
18-27 °C
H
24 in. (61 cm)
610 mm
General description
These fascinating and functional instruments indicate temperature by colorful glass spheres that rise and fall according to changes in temperature of the clear liquid contained in each unit. Based on Galileo Galilei′s discovery that as a liquid′s temperature increases, its buoyancy decreases.
• Calibrated multi-colored glass spheres, in °F or °C
• The lowest floating sphere indicates temperature
• All-glass, hand blown body filled with harmless clear liquid
Choose from one of the most recent versions:
Certificates of Analysis (COA)