Key Documents
Z309281
Quickfit® flat-bottom boiling flasks
capacity 100 mL, joint: ST/NS 19/26
Synonym(s):
FF100/2S
Select a Size
1 EA
$76.50
5 EA
$236.00
$76.50
Estimated to ship on09 April 2025
Select a Size
Change View
1 EA
$76.50
5 EA
$236.00
About This Item
UNSPSC Code:
41121804
$76.50
Estimated to ship on09 April 2025
material
borosilicate glass
flat bottom flask
short
Joint
joint: ST/NS19/26
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 5
manufacturer/tradename
Quickfit FF 100/2S
availability
not available in USA and Canada
capacity
100 mL
General description
With short neck and ground-glass socket. Not suitable for vacuum use.
Features and Benefits
• Complies with ISO 4797 and ISO 1773• Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)• The flat base allows use with hotplates• FF150/3S, FF150/4S, FF150/5S and FF500/2S are supplementary to ISO 4797.• Note: These flasks are not suitable for use under a positive or negative pressure
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)