Key Documents
Z309265
Quickfit® flat-bottom boiling flasks
capacity 50 mL, joint: ST/NS 19/26
Synonym(s):
FF50/2S
Select a Size
5 EA
$226.00
$226.00
Check Cart for Availability
Select a Size
Change View
5 EA
$226.00
About This Item
UNSPSC Code:
41121804
$226.00
Check Cart for Availability
material
borosilicate glass
flat bottom flask
short
Joint
joint: ST/NS19/26
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 5
manufacturer/tradename
Quickfit FF 50/2S
availability
not available in USA and Canada
capacity
50 mL
General description
With short neck and ground-glass socket. Not suitable for vacuum use.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)