Key Documents
Z309176
Quickfit® Soxhlet extractor body
volume 100 mL
Synonym(s):
EX5/63
Select a Size
1 EA
$313.00
$313.00
Check Cart for Availability
Select a Size
Change View
1 EA
$313.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
$313.00
Check Cart for Availability
manufacturer/tradename
Quickfit EX5/63
availability
not available in USA and Canada
volume measuring range
100 mL
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Hi, may I know what is the top and bottom socket size for this item?
    1 answer
    1. The bottom cone size is 24/29 and the top socket size is 40/38.
      Helpful?