Skip to Content
Merck
All Photos(1)

Key Documents

Z308536

Quickfit® sintered bottle heads

for use with 500 mL bottles

Synonym(s):

MF48/13/500

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$247.00

$247.00


Check Cart for Availability


Select a Size

Change View
1 EA
$247.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.51

$247.00


Check Cart for Availability

sterility

non-sterile

manufacturer/tradename

Quickfit MF 48/13/500

availability

not available in USA and Canada

compatibility

for use with 500 mL bottles

Looking for similar products? Visit Product Comparison Guide

General description

Sintered heads improve gas dispersion

Legal Information

Quickfit is a registered trademark of SciLabware Limited

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service