Key Documents
Z304662
Quickfit® ground-glass stoppers
joint: ST/NS 45/40
Synonym(s):
SB45
Select a Size
1 EA
$116.00
$116.00
Check Cart for Availability
Select a Size
Change View
1 EA
$116.00
About This Item
UNSPSC Code:
41122500
NACRES:
NB.35
$116.00
Check Cart for Availability
Joint
joint: ST/NS45/40
manufacturer/tradename
Quickfit SB 45
availability
not available in USA and Canada
General description
  • Ideally suited for use with Quickfit® jointedware and other glassware having standard ground glass sockets
  • All Quickfit® stoppers conform to the standard tapers as outlined in ISO 383
  • Borosilicate glass for chemical durability and high temperature performance
  • Hollow-blown construction to reduce weight
  • Hexagonal head prevents the stopper from rolling and provides a good grip
  • Top of stopper is flat, allowing them to be stood on end and avoid contamination of the ground surface
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)