Skip to Content
Merck
All Photos(1)

Key Documents

Z303321

Quickfit® jacketed coil condenser

L 160 mm, joint: ST/NS 29/32

Synonym(s):

CX6/44/SC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

Joint

joint: ST/NS29/32

manufacturer/tradename

Quickfit CX 6/44/SC

availability

not available in USA and Canada
not available in USA and Canada

L

160 mm

surface area

400 cm2

Looking for similar products? Visit Product Comparison Guide

General description

Double surface condensers with a coil as the inner surface provides approximately twice the surface area of plain, double surface condensers of the same effective length.

Legal Information

Quickfit is a registered trademark of SciLabware Limited

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service