Key Documents
Z301094
Quickfit® expansion adapter
top female joint: ST/NS 19/26, bottom male joint: ST/NS 14/23
Synonym(s):
XA21
Select a Size
1 EA
$72.80
$72.80
Available to ship on22 April 2025Details
Select a Size
Change View
1 EA
$72.80
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$72.80
Available to ship on22 April 2025Details
Joint
female joint: ST/NS top19/26
male joint: ST/NS bottom14/23
manufacturer/tradename
Quickfit XA 21
availability
not available in USA and Canada
not available in USA and Canada
General description
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)