Key Documents
Z285498
Sigma-Aldrich
DigiTrol II temperature controller
AC/DC input 120 V AC, 15 A
Synonym(s):
Temperature controller, Temperature monitoring system
About This Item
UNSPSC Code:
41112303
NACRES:
NB.76
Product Z285498 is not currently sold in your country. Contact Technical Service
manufacturer/tradename
Glas-Col 104APL612K
availability
not available in EU
parameter
32-999 °F temperature (0-750 °C)
output
1,800 watts
General description
Simultaneously displays set point and bath temperature to 1.0°C. Supplied with a Type K, 6 in. immersion thermocouple probe that fits Aldrich oil baths. 240 V models are supplied with a US connector.
Choose from one of the most recent versions:
Certificates of Analysis (COA)