Key Documents
Z281239
Anti-static brush
ideal for balance and lens cleaning
Synonym(s):
Anti-Static Cleaning Brush
About This Item
UNSPSC Code:
47131605
NACRES:
NB.35
Product Z281239 is not currently sold in your country. Contact Technical Service
W × L
66 mm × 63 mm
General description
This anti-static wisk style cleaning brush is particularly suitable for cleaning sensitive surfaces and at the same time removing any existing static charge.
Ideal for balance and lens cleaning without any danger to the coating. The brush consists of a mixture of highly-flexible natural fibers and conductive synthetic fibers as well as a handle made of conductive plastic.
After they have been cleaned the treated surfaces are electrically neutral, and do not attract any dust particles. Dirty brushes can be washed without this influencing their anti-static effect. The brushes can be used at temperatures of up to 65°.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Questions
  1. Hi, Could you advise what is the brush wire made of? * Z281239 Anti-static brush I need a Conductive RUBBER Brush. Thank you
    1 answer
    1. The unique blend of soft natural hairs and conductive synthetic fibers plus the conductive nature of the construction result in effective static dissipation that removes static as it cleans.
      Helpful?