Skip to Content
Merck
All Photos(1)

Key Documents

Z2753

Sigma-Aldrich

trans-Zeatin hydrochloride

suitable for plant cell culture, ≥97%

Synonym(s):

6-(4-Hydroxy-3-methylbut-2-enylamino)purine, N6-(4-Hydroxy-3-methyl-2-buten-1-yl)adenine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$180.00

$180.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$180.00

About This Item

Empirical Formula (Hill Notation):
C10H13N5O · HCl
CAS Number:
Molecular Weight:
255.70
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$180.00


Check Cart for Availability

Request a Bulk Order

Assay

≥97%

technique(s)

cell culture | plant: suitable

solubility

H2O: soluble

application(s)

agriculture

storage temp.

−20°C

SMILES string

C\C(CO)=C/CNc1ncnc2[nH]cnc12

InChI

1S/C10H13N5O/c1-7(4-16)2-3-11-9-8-10(13-5-12-8)15-6-14-9/h2,5-6,16H,3-4H2,1H3,(H2,11,12,13,14,15)/b7-2+

InChI key

UZKQTCBAMSWPJD-FARCUNLSSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marek Zatloukal et al.
Bioorganic & medicinal chemistry, 16(20), 9268-9275 (2008-09-27)
The synthesis of a new group of 2-X-6-anilinopurines, including compounds with potential cytokinin-like activities, with various substitutions (X=H, halogen, amino, methylthio or nitro) on the phenyl ring is described. The prepared compounds have been characterized using standard physico-chemical methods, and
Isaiah Taylor et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(8) (2021-02-21)
Early root growth is critical for plant establishment and survival. We have identified a molecular pathway required for helical root tip movement known as circumnutation. Here, we report a multiscale investigation of the regulation and function of this phenomenon. We
Lucie Szücová et al.
Bioorganic & medicinal chemistry, 17(5), 1938-1947 (2009-02-24)
In an attempt to improve specific biological functions of cytokinins routinely used in plant micropropagation, 33 6-benzylamino-9-tetrahydropyran-2-ylpurine (THPP) and 9-tetrahydrofuran-2-ylpurine (THFP) derivatives, with variously positioned hydroxy and methoxy functional groups on the benzyl ring, were prepared. The new derivatives were
Jonas Van Bockhaven et al.
The New phytologist, 206(2), 761-773 (2015-01-28)
Although numerous studies have shown the ability of silicon (Si) to mitigate a wide variety of abiotic and biotic stresses, relatively little is known about the underlying mechanism(s). Here, we have investigated the role of hormone defense pathways in Si-induced
Fuyou Du et al.
Analytical and bioanalytical chemistry, 407(20), 6071-6079 (2015-05-31)
High-internal-phase-emulsion polymers (polyHIPEs) show great promise as solid-phase-extraction (SPE) materials because of the tremendous porosity and highly interconnected framework afforded by the high-internal-phase-emulsion (HIPE) technique. In this work, polyHIPE monolithic columns as novel SPE materials were prepared and applied to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service