Key Documents
Z268771
Safe-Lab Thermometer/Tubing Adapter
joint: ST/NS 10/18, opening size 6-7 mm
Synonym(s):
thermometer holder
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
joint: ST/NS10/18
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F20309-1675
opening size
6-7 mm
General description
PTFE body with Viton® O-ring and PP screw-cap with inside Viton O-ring for air-tight work. Loosen cap for depth adjustment of thermometer or tubing.
Legal Information
Safe-Lab is a trademark of SP Scienceware, a division of SP
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)