Key Documents
Z268755
Scienceware® Safe-Lab PTFE stirrer bearing
joint: ST/NS 29/42
Select a Size
1 EA
$308.00
$308.00
Estimated to ship on09 April 2025
Select a Size
Change View
1 EA
$308.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
$308.00
Estimated to ship on09 April 2025
material
Viton® O-ring
Joint
joint: ST/NS29/42
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F20320-1662
General description
Stopper-Loc® nut allows stirrer bearing to be locked to outer ground-glass joint. Complete with Stopper-Loc nut and two PTFE inserts. For 10mm diam. polished glass or PTFE stirring shafts.
Legal Information
Safe-Lab is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Stopper-Loc is a registered trademark of E. I. du Pont de Nemours and Company
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)