Z262803
Wheaton narrow mouth bottles with caps
capacity 2 oz (60 mL), clear, black plastic cap, Poly-Seal® cone liner
Synonym(s):
Wheaton bottle, Wheaton bottles, narrow mouth
Select a Size
$146.00
Select a Size
About This Item
$146.00
Recommended Products
material
black plastic cap
clear bottle
glass bottle
round bottle
Joint
Joints threaded neck (20-400)
feature
with cap
packaging
pkg of 24 ea
manufacturer/tradename
WHEATON® W216813
capacity
2 oz (60 mL)
diam. × H
39 mm × 94 mm
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Choice of cap styles:
- White PP cap with PTFE or polyvinyl liner
- Black plastic cap with Poly-Seal® PE cone liner
Features and Benefits
- Manufactured from clear USP Type III soda-lime glass
- Polypropylene caps with choice of liners
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service