Z252379
Aldrich® anticlimb splash-guard adapter with fritted disc
female joint: ST/NS 29/32, male joint: ST/NS 29/32, capacity 500 mL
All Photos(1)
About This Item
Recommended Products
Joint
female joint: ST/NS29/32
male joint: ST/NS29/32
capacity
500 mL
pore size
70-100 μm porosity
Related Categories
General description
These traps greatly reduce carry-over from foaming or bumping. Because the venting hole is located at the top, the foam can be prevented from climbing higher into the condenser. The fritted disc prevents particulates from entering the condenser.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC