Key Documents
Z252352
Sigma-Aldrich
Aldrich® anticlimb splash-guard adapter with fritted disc
female joint: ST/NS 29/32, male joint: ST/NS 29/32, capacity 100 mL
Select a Size
1 EA
$286.00
$286.00
Check Cart for Availability
Select a Size
Change View
1 EA
$286.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$286.00
Check Cart for Availability
Joint
female joint: ST/NS29/32
male joint: ST/NS29/32
capacity
100 mL
pore size
70-100 μm porosity
Related Categories
General description
These traps greatly reduce carry-over from foaming or bumping. Because the venting hole is located at the top, the foam can be prevented from climbing higher into the condenser. The fritted disc prevents particulates from entering the condenser.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)