Key Documents
Z247235
Coors porcelain crucible cover
cap size D, I.D. 54 mm
Select a Size
1 PKG
$381.00
$381.00
Check Cart for Availability
Select a Size
Change View
1 PKG
$381.00
About This Item
UNSPSC Code:
41122200
NACRES:
NB.43
$381.00
Check Cart for Availability
cap size
D
packaging
pkg of 12 ea
manufacturer/tradename
Coorstek 66125
I.D.
54 mm
General description
Coors porcelain crucible cover is glazed all over except for the rim.
Features and Benefits
  • Ensures great resistance to thermal-shock
  • Durable
  • Rigid
  • Sustained refractory qualities
Other Notes
Covers may not fit exactly on crucibles due to unavoidable wall thickness variations within the crucible.
Legal Information
Coors is a trademark of Coorstek, Inc.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Kindly describe how many pieces of lid are there in this pack.
    1 answer
    1. This product is sold as a package of 12 each. There are 12 covers included.
      Helpful?