Key Documents
Z217972
Supelco
Michel-Miller column coupling to end fittings
size 15 mm
About This Item
UNSPSC Code:
23151816
manufacturer/tradename
Ace Glass 584047
technique(s)
LPLC: suitable
size
15 mm
General description
PTFE; for coupling column to end fittings of the same internal diameter. Size refers to inside diameter of threads. Includes two FETFE® O-rings.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)