Key Documents
Z215511
Rigid PTFE sleeve
joint: ST/NS 29/32
Synonym(s):
ptfe sleeves
Select a Size
1 EA
$39.40
$39.40
Check Cart for Availability
Select a Size
Change View
1 EA
$39.40
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
$39.40
Check Cart for Availability
Joint
joint: ST/NS29/32
manufacturer/tradename
Cowie
General description
Robust, PTFE sleeve with substantial grip-ring for repeated use and ease of handling. Transverse trapezoidal sealing rings provide grease-free, high-vacuum performance at temperatures up to 280°C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)