Z204005
Flow controllers for flash-chromatography assemblies with ball joints
joint: SJ 29/15
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121813
NACRES:
NC.05
Joint
joint: SJ29/15
grade
for chromatography
technique(s)
LPLC: suitable
suitability
suitable for chromatography
compatibility
for use with 29/15 standard ball joint
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service