Key Documents
Z190403
Sigma-Aldrich
Aldrich® flask-type sprayer
Replacement Head only
Select a Size
1 EA
$256.00
$256.00
Check Cart for Availability
Select a Size
Change View
1 EA
$256.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$256.00
Check Cart for Availability
material
borosilicate glass
sterility
non-sterile
feature
closure type Rubber cork
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
Related Categories
General description
Connect using the barb end to a low-pressure gas or air source of <5 psi. By positioning your thumb over the vent hole you can control the spray pattern.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)