Skip to Content
Merck
All Photos(1)

Key Documents

Z186848

Sigma-Aldrich

All-purpose sampling containers

capacity 8 oz

Sign Into View Organizational & Contract Pricing

Select a Size

500 EA
$330.00

$330.00


Available to ship on22 April 2025Details



Select a Size

Change View
500 EA
$330.00

About This Item

UNSPSC Code:
41105108
NACRES:
NB.51

$330.00


Available to ship on22 April 2025Details


capacity

8 oz

Looking for similar products? Visit Product Comparison Guide

General description

Durable and translucent PP containers resist acids, bases, most organic solvents, and common laboratory reagents. Internally molded dual graduations in increments of 1 oz and 20 mL. Economical enough to discard after one use, or strong enough to reuse. Tight-fitting PE lid not included. Nonsterile.

Other Notes

Supplied without lid. No lid available.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service