Key Documents
Z179825
Supelco
Michel-Miller column stopper
Ace-Thred for #25
Select a Size
1 EA
$202.00
$202.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$202.00
About This Item
UNSPSC Code:
12000000
NACRES:
SB.52
$202.00
Check Cart for Availability
Request a Bulk Order
manufacturer/tradename
Ace Glass 580325
technique(s)
LPLC: suitable
fitting
Ace-Thred for #25
General description
PTFE for use at either end of any chromatography column with Ace-Threds. Makes a leak-tight seal without O-rings.
Choose from one of the most recent versions:
Certificates of Analysis (COA)