Z167290
Suba-Seal® septa
white rubber, Suba-Seal® 33, fits neck I.D., 17.5 mm, for use with all Sure/Seal bottles
Select a Size
$284.00
Select a Size
About This Item
$284.00
Recommended Products
material
white rubber
feature
fits neck I.D. 17.5 mm
for use with all Sure/Seal bottles
manufacturer/tradename
Suba-Seal® 33
Looking for similar products? Visit Product Comparison Guide
General description
This closure has turnover flange molded to grip outside of container neck promoting a double seal in conjunction with annular serrations on hollow plug. Annular serrations depress themselves against the inside wall of the bottle neck, making each serration a suctions sealing point.
Storage Note for Natural Rubber Septa: Store septa in a closed bag out of sunlight to retard blooming, which is the formation of a whitish film on the surface of the rubber. Blooming is a natural process and does not affect performance. Remove visible bloom by washing or wiping off before use.
Note: For extra security when using large bore transfer needles, fasten septum to bottle or flask opening with a wrap-it tie. See ties and accessory tool listings.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service