Key Documents
Z156515
Sigma-Aldrich
Pinch clamps for SJ joints
joint: SJ 28/12
Select a Size
1 EA
$75.60
$75.60
Check Cart for Availability
Select a Size
Change View
1 EA
$75.60
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
$75.60
Check Cart for Availability
material
stainless steel
Joint
joint: SJ28/12
General description
Use with flash-chromatography ball jointed assemblies and rotary evaporator receiver flasks.
Choose from one of the most recent versions:
Certificates of Analysis (COA)