Z153915
Duran® plastic-coated laboratory bottles
capacity 2,000 mL
Synonym(s):
bottle laboratory, lab bottle
About This Item
Recommended Products
material
borosilicate glass 3.3
clear bottle
narrow-mouth bottle
round bottle
feature
autoclavable
without cap
packaging
pack of 1 ea
pack of 10 ea
manufacturer/tradename
DURAN® 21 805 63
parameter
max. pressure (Not for use under pressure)
capacity
2,000 mL
diam. × H
136 mm × 260 mm
fitting
GL 45 neck thread
Looking for similar products? Visit Product Comparison Guide
General description
Please note: Cap not included.
Features and Benefits
- Made from borosilicate glass 3.3 DURAN®
- According to USP <660>, EP (3.2.1), and JP <7.01>, DURAN® is a neutral glass that is categorized as a Glass Type I and complies with ASTM E438, Type I, Class A requirements.
- The external polymer coating offers protection against scratches, leaks, and splinters (only for bottles containing 2000 ml or less).
- UV protection up to approximately 380 nm due to external coating
- High resistance and long-lasting coating
- Standardized GL-thread and matching cap system; for mainly tight sealing and simple, clean pouring out
- Fully equipped with a screw cap (PP), integral lip seal, and pouring ring (PP), allowing for drip-free pouring and clean, safe working.
- Sizes 10 and 25 ml with specially shaped glass edges for improved pouring out, preventing the need for an additional plastic pouring ring
- Fired-on, highly durable white ceramic print
Legal Information
required but not provided
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service