Key Documents
Z147338
Sigma-Aldrich
Lecture-bottle adapter, NPT to NPT
stainless steel
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
material
stainless steel
inlet and outlet NPTM × NPTM
1/4 in. × 1/4 in.
General description
For connection of valves, regulators, and filters to cylinder valves.
Choose from one of the most recent versions:
Certificates of Analysis (COA)