Key Documents
Z137383
Ace Trubore® 3-piece glass stirrer bearing
joint: ST/NS 19/22, fits shaft o.d., 6 mm
Select a Size
1 EA
$239.00
$239.00
Check Cart for Availability
Select a Size
Change View
1 EA
$239.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
$239.00
Check Cart for Availability
Joint
joint: ST/NS19/22
feature
fits shaft o.d. 6 mm
manufacturer/tradename
Ace Glass 952910
Related Categories
General description
Self-aligning, three-piece bearing consisting of a glass inner Trubore bearing, nylon bushing with O-ring, and threaded glass adapter. Inner bearing will spin if shaft binds or will self-align at O-ring if motor is slightly cocked. Inner bearing and bushing can be used with any joint size.
Legal Information
Trubore is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)