Key Documents
Z125520
Ace-Thred Thermometer/Tubing Adapter
With hose connection, joint: ST/NS 19/22, thread I.D. 7.0 mm
Select a Size
1 EA
$103.00
$103.00
Check Cart for Availability
Select a Size
Change View
1 EA
$103.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$103.00
Check Cart for Availability
label
F
Joint
joint: ST/NS19/22
manufacturer/tradename
Ace Glass 526112
thread I.D.
7.0 mm
Related Categories
General description
Threaded nylon bushing with FETFE® O-ring.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
FETFE is a registered trademark of Ace Glass, Inc.
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)