Key Documents
Z118982
Sigma-Aldrich
Scienceware® Frigimat® dry ice makers
USA connectors
Select a Size
1 EA
$2,610.00
$2,610.00
Check Cart for Availability
Select a Size
Change View
1 EA
$2,610.00
About This Item
UNSPSC Code:
41103000
$2,610.00
Check Cart for Availability
material
aluminum ice chamber
manufacturer/tradename
Bel-Art
availability
not available in EU
General description
A 6ft nickel plated copper connecting tube with USA standard CGA 320 inlet for attachment to CO2 cylinder is included.
Produces solid 1 lb cylindrical block 8.5 × 11 cm of dry ice from a cylinder of liquid CO2. One CO2 cylinder makes 8 to 9 blocks of dry ice. Epoxy-coated steel frame, brass valve, aluminum dry ice chamber.
Legal Information
Frigimat is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)