Key Documents
Z117080
Stainless steel 304 syringe needle
gauge 16, L 10 in.
Select a Size
1 EA
$55.30
$55.30
Check Cart for Availability
Select a Size
Change View
1 EA
$55.30
About This Item
UNSPSC Code:
41122002
eCl@ss:
32070500
NACRES:
NB.03
$55.30
Check Cart for Availability
description
Standard Hub with 304 Stainless Steel Tubing
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science
L
10 in.
gauge
16
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Choose from one of the most recent versions:
Certificates of Analysis (COA)