Key Documents
Z105805
Polyethylene stopper
joint: ST/NS 19/38
Synonym(s):
flask stopper, flask stoppers, stoppers
Select a Size
100 EA
$35.40
$35.40
Available to ship on28 April 2025Details
Select a Size
Change View
100 EA
$35.40
About This Item
UNSPSC Code:
41122500
NACRES:
NB.35
$35.40
Available to ship on28 April 2025Details
Joint
joint: ST/NS19/38
General description
Nonthreaded, wide, thick flanges.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. HI, Can you provide dimensions of these stoppers. Ineed a stopper that will plug a hole 22mm in diameter
    1 answer
    1. Please see the image below to review the dimensions.
      Helpful?