Key Documents
Z100617
Sigma-Aldrich
Single-bulb ball-tube
capacity 500 mL, joint: ST/NS 14/20, plain
Select a Size
1 EA
$209.00
$209.00
Check Cart for Availability
Select a Size
Change View
1 EA
$209.00
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
$209.00
Check Cart for Availability
Joint
joint: ST/NS14/20
capacity
500 mL
General description
Arranged horizontally outside of Kugelrohr oven, the ball-tubes cool and collect distillate with minimal loss of material.
Choose from one of the most recent versions:
Certificates of Analysis (COA)