Key Documents
WHAMUPMCPBC8
Whatman® Mini-UniPrep® syringeless filters accessories
Multicompressor unit
Select a Size
1 EACH
$580.00
$580.00
Check Cart for Availability
Select a Size
Change View
1 EACH
$580.00
About This Item
UNSPSC Code:
41104922
NACRES:
NB.24
$580.00
Check Cart for Availability
sterility
non-sterile
feature
Multicompressor unit
manufacturer/tradename
Whatman MUPMCPBC8
General description
Contains a Multicompressor and a single 8 position blue tray to be used with plastic MUP
Features and Benefits
  • Streamlines syringeless filtration by filtering up to eight filter vials simultaneously
  • Designed for Cytiva′s Mini-UniPrepstandard plastic filter vial
  • Mini-UniPrepvials can be loaded directly into the autosampler
Legal Information
Uniprep is a registered trademark of Whatman International Ltd.
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)