WHA69702504
Whatman® GD/XP syringe filters
Nylon membrane
Synonym(s):
Whatman filter, Z338389, syringe filter, syringe filters
Select a Size
$713.00
Available to ship on29 April 2025Details
Select a Size
About This Item
$713.00
Available to ship on29 April 2025Details
Recommended Products
material
Nylon membrane
polypropylene housing
sterility
non-sterile
feature
Polypropylene prefilter
packaging
pack of 150 ea
manufacturer/tradename
Whatman 6970-2504
Whatman Article No. 28420561 (US reference)
diam.
25 mm
volume
≤25 mL
pore size
0.45 μm
fitting
female Luer inlet (lock)
male Luer outlet
Features and Benefits
- Incorporates a two-layer polypropylene prefilter stack rather than a multi-layer glass fiber stack to minimize levels of ion extractables
- Extremely low protein binding syringe filter
- High loading volume for filtration of challenging samples
Suitability
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)