WHA5226200
Whatman® filter paper, Grade 226, creped, grey
circles, diam. 200 mm (7.9 in.), pkg of 100 ea
Synonym(s):
Whatman filter, Whatman paper, Z761877, paper filter
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
circles
creped gray cellulose
packaging
pkg of 100 ea
manufacturer/tradename
Whatman 5226-200
parameter
fast flow rate
fast flow rate
diam.
200 mm (7.9 in.)
thickness
0.28 mm (0.01 in.)
pore size
20-25 μm
basis weight
87 g/m2
capacity
high-normal loading
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Filter papers are manufactured primarily for non-critical industrial laboratory filtration applications requiring inexpensive filters of consistent performance.
Grade 226 is a unique filter paper with a creped, grey surface providing optical contrast for white or dark filtrates. Retains coarse or gelatinous precipitates. Filtration speed is high. Grade 226 is used extensively in sugar laboratories for routine and/or repetitive filtration.
Note; These papers should not be used for kjeldahl nitrogen analysis.
Grade 226 is a unique filter paper with a creped, grey surface providing optical contrast for white or dark filtrates. Retains coarse or gelatinous precipitates. Filtration speed is high. Grade 226 is used extensively in sugar laboratories for routine and/or repetitive filtration.
Note; These papers should not be used for kjeldahl nitrogen analysis.
Features and Benefits
- High filtration speed
- Retains coarse or gelatinous particulates
- Unique crêped gray surface provides optical contrast for white or dark filtrates
Legal Information
Whatman is a registered trademark of Cytiva
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service