WHA10443001
Whatman® glass vacuum filtration device
GV100 series, GV100/0/01 funnel, 500 mL
Synonym(s):
Z750913
Select a Size
$318.00
Select a Size
About This Item
$318.00
Recommended Products
material
borosilicate glass funnel(s) (funnel)
borosilicate glass housing
stainless steel clamp(s) (clamp)
packaging
pk of 1
manufacturer/tradename
Whatman 10443001
Whatman Article No. 28415575 (US reference)
filter capacity
500 mL
funnel capacity
500 mL
suitability
suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions)
Related Categories
General description
Produced from borosilicate glass and available with a choice of support screen. Suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
The sintered glass support is recommended for filtration and biological analysis. The 304 stainless steel support screen is suitable for use with proteinaceous solutions.
Features and Benefits
- Accessory to GV100 Glass Filter Funnel
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)