Skip to Content
Merck
All Photos(2)

Key Documents

WHA10402012

Whatman® nitrocellulose NC10 filter discs 0.1 μm pore size

white, hydrophilic, 100 ea, 47 mm diam

Synonym(s):

Whatman filter, Z697370, cellulose membrane, membrane filter

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$294.00

$294.00


Check Cart for Availability


Select a Size

Change View
1 PKG
$294.00

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24

$294.00


Check Cart for Availability

Product Name

Whatman® nitrocellulose membrane filters, NC10 Membrane Circles, diam. 47 mm, pore size 0.1 μm

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Whatman 10402012
Whatman Article No. 28415055 (US reference)

packaging

pkg of

diam.

47 mm

pore size

0.1 μm

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Standard membrane filter for most applications involving particles and cells from 0.1 µm to 12.0 µm
  • Samples are retained on the surface for easy microscopic analysis and recovery
  • Temperature-resistant cellulose nitrate can withstand autoclaving up to 121°C with no loss in integrity
  • Whatman membrane filters exhibit a low shrinkage during autoclaving, reducing tearing
  • High strength and flexibility prevent tearing during handling, loading, and autoclaving

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dori M Landry et al.
Genes & development, 23(23), 2753-2764 (2009-12-03)
Most eukaryotic mRNAs are translated using a cap-dependent mechanism of translation. However, approximately 10% of mammalian mRNAs initiate translation using a cap-independent mechanism that is not well understood. These mRNAs contain an internal ribosome entry site (IRES) located in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service