WHA10370305
Whatman® glass microfiber grade GF 10 filter discs
white, organic binder, 100 ea, 90 mm diam
Synonym(s):
Z749982
Select a Size
$232.00
Select a Size
About This Item
$232.00
Recommended Products
Product Name
Whatman® glass microfiber filters with organic binder, Grade GF 10, circles, diam. 90 mm, pack of 100
material
circles
glass (with inorganic binder)
feature
hydrophilic
packaging
pack of 100
manufacturer/tradename
Whatman 10370305
Whatman Article No. 28414782 (US reference)
diam.
90 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Temperature resistance up to 180°C
- Strengthened with high-quality organic binder for high mechanical stability
- Medium filtration speed
- Can be used as a roll filter in automated applications
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service