WHA10314726
Whatman® qualitative filter paper, Grade 1573
circles, diam. 290 mm, pack of 100
Synonym(s):
Z749370
Select a Size
$203.00
Select a Size
About This Item
$203.00
Recommended Products
material
cellulose filters
circles
packaging
pack of 100
manufacturer/tradename
Whatman 10314726
Whatman Article No. 28414345 (US reference)
parameter
25 sec/100 mL speed (Herzberg)
diam.
290 mm
thickness
170 μm
pore size
12-25 μm (Particle retention)
basis weight
88 g/m2
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Grade 1573: 12-25 μm (fast, smooth filter paper)
A fast filter paper with high wet strength. It has a very smooth surface, making it easy to scrape or wash off precipitate. Resistant against: sulfuric and nitric acid solutions (up to 40% at 50°C), hydrochloric (up to 10% at 100°C, 20% at 60°C, 25% at 20°C), alkalis (up to 10% at 20°C).
Features and Benefits
- A Fast filter paper with high wet strength. It has a very smooth surface making it easy to scrape or wash off precipitate.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service