Skip to Content
Merck
All Photos(1)

Key Documents

WHA1030024

Whatman® cellulose chromatography papers

3 mm blotting paper, diam. 24 mm

Synonym(s):

Z752428

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$23.90

$23.90


Check Cart for Availability


Select a Size

Change View
1 PKG
$23.90

About This Item

UNSPSC Code:
41115727
NACRES:
NB.24

$23.90


Check Cart for Availability

material

cellulose membrane
circles

sterility

non-sterile

packaging

pkg of 100 ea

manufacturer/tradename

Whatman 1030-024
Whatman Article No. 28414094 (US reference)

parameter

1385 sec / 100 mL speed

diam.

24 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Grade 3MM Chr cellulose chromatography circles is a 0.34 mm paper for general chromatography and electrophoresis. Though widely used as a blotting paper, 3MM Chr is also used both in electrophoresis and for general chemistry. A medium thickness paper (0.34 mm) used extensively for general chromatography and electrophoresis. Flow rate is 130 mm/30 min.

Features and Benefits

  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind. Ensures that no contamination will occur during the transfer steps.
  • Manufactured and tested specifically for chromatographic and blotting techniques. This ensures the wicking capability and uniformity of capillary action that are important in obtaining clean and even transfers during blotting.
  • Whatman 3MM Chr is considered the industry standard for blotting procedures.
  • Convenient sizes available in sheets precisely cut to the most popular gel and transfer membrane sizes. Allows "out-of-the-box" usage and eliminates sheet-to-sheet variations.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susana Campuzano et al.
Applied and environmental microbiology, 75(18), 5972-5980 (2009-07-28)
A Streptococcus mitis genomic DNA fragment carrying the SMT1224 gene encoding a putative beta-galactosidase was identified, cloned, and expressed in Escherichia coli. This gene encodes a protein 2,411 amino acids long with a predicted molecular mass of 268 kDa. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service