Skip to Content
Merck
All Photos(2)

Key Documents

W800411

Sigma-Aldrich

Methyl cyclomyrcetone

mixture of isomers, ≥90%

Synonym(s):

7-Acetyl-1,2,3,4,5,6,7,8-octahydro-1,1,6,7-tetramethylnaphthalene, Patchouli ethanone, 1-(1,2,3,4,5,6,7,8-Octahydro-2,3,8,8-tetramethyl-2-naphthalenyl)ethanone, 2-Acetyl-2,3,8,8-tetramethyloctalin, Iso E Super

Sign Into View Organizational & Contract Pricing

Select a Size

W800411-SAMPLE
$75.30
1 KG
$121.00
10 KG
$565.00

$75.30


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W800411-SAMPLE
$75.30
1 KG
$121.00
10 KG
$565.00

About This Item

Empirical Formula (Hill Notation):
C16H26O
CAS Number:
Molecular Weight:
234.38
Beilstein:
1961701
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
Fragrance grade
Halal
food allergen:
no known allergens

$75.30


Check Cart for Availability

Request a Bulk OrderRequest more information

grade

Fragrance grade
Halal

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

≥90%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

cedar; woody; phenolic

SMILES string

O=C(C1(C(CC2=C(C(CCC2)(C)C)C1)C)C)C

InChI

1S/C16H26O/c1-11-9-13-7-6-8-15(3,4)14(13)10-16(11,5)12(2)17/h11H,6-10H2,1-5H3

InChI key

FVUGZKDGWGKCFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Resp. Sens. 1B - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service