Skip to Content
Merck
All Photos(1)

Key Documents

W530293

Sigma-Aldrich

Jojoba oil

Certified organic (NOP)

Synonym(s):

Jojoba oil from Simmondsia chinensis, Jojoba bean oil, Jojoba liquid wax

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164502
grade:
Certified organic (NOP)
Kosher
food allergen:
no known allergens

grade

Certified organic (NOP)
Kosher

origin

USA origin

refractive index

n20/D 1.466

density

0.868 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

tropical

Looking for similar products? Visit Product Comparison Guide

General description

Jojoba oil is a liquid wax obtained from the seeds of jojoba plant (Simmondsia chinensis). It is generally composed of long chain esters of fatty acids and fatty alcohols. Its minor constituents may include free fatty acids and alcohols, phytosterols, tocopherols phospholipids, and triglycerides.[1] The fatty acid methyl esters (FAME) produced by the transesterification of jojoba oil-wax with methanol shows promising potential as biodiesel.[2][3]

Legal Information

Certified organic by MOSA

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extraction of jojoba oil by pressing and leaching.
Abu-Arabi MK, et al.
Chemical Engineering Journal, 76(1), 61-65 (2000)
Structural determination and uses of jojoba oil.
Miwa TK
Journal of the American Oil Chemists' Society, 61(2), 407-410 (1984)
Biodiesel from Jojoba oil-wax: Transesterification with methanol and properties as a fuel.
Canoira L, et al.
Biomass and Bioenergy, 30(1), 76-81 (2006)
Elia Ranzato et al.
Journal of ethnopharmacology, 134(2), 443-449 (2011-01-08)
The wound healing properties of jojoba (Simmondsia chinensis) liquid wax (JLW) were studied in vitro on HaCaT keratinocytes and human dermal fibroblasts, which are involved in wounded skin repair. JLW cytotoxicity was evaluated by the crystal violet staining and the
Tania Cristina de Sá Dias et al.
Journal of cosmetic dermatology, 7(2), 120-126 (2008-05-17)
Straightening is a chemical process by which excessively curly hair is straightened in an irreversible way. Generally, products are formulated as emulsions with high pH value (9.0-12.0), which, after applied on hair, cause considerable damage, making it dry and fragile.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service